Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990126
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0126 25 μL
50-199-0125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0126 Supplier Rockland Immunochemicals Supplier No. 100401GN3S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Glut-2 antibody is directed against Glut2 protein. A BLAST analysis was used to suggest cross-reactivity with Glut2 from mouse and rat based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.

Specifications

Antigen Glut2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Slc2a2
Gene Accession No. P11168, P12336, P14246
Gene Alias AI266973; Glucose transporter type 2, liver; Glut2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; Solute carrier family 2, facilitated glucose transporter member 2
Gene Symbols Slc2a2
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminal domain of mouse Glut2 protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20526, 25351, 6514
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.