Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980228
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0228 Supplier Bioss Supplier No. BS0351R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glucose transporter 2 isoform is an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. It mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in the gene that encode this protein cause Fanconi-Bickel syndrome.

Specifications

Antigen GLUT2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Slc2a2
Gene Accession No. P11168
Gene Alias AI266973; Glucose transporter type 2, liver; Glut2; GLUT-2; GTT2; liver-type glucose transporter; Slc2a2; solute carrier family 2 (facilitated glucose transporter), member 2; solute carrier family 2 member 2; Solute carrier family 2, facilitated glucose transporter member 2
Gene Symbols Slc2a2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GLUT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6514
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.