Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596952
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596952 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596952 Supplier Invitrogen™ Supplier No. PA596952
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESPRFLLINR KEEENAKQIL QRLWGTQDVS QDIQEMKDES ARMSQEKQVT VLELFRVSSY RQPIIISIVL QLSQQ Positive Samples: HeLa, HepG2, U-87MG; Cellular Location: Cell membrane, Multi-pass membrane protein.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC2A3
Gene Accession No. P11169, P32037, Q07647
Gene Alias AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3
Gene Symbols SLC2A3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 207-281 of human GLUT3/SLC2A3 (NP_0088621).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20527, 25551, 6515
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.