Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUT3 Polyclonal Antibody
GLUT3 Polyclonal Antibody
Supplier: Thermo Scientific PA599486
Description
GLUT3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
GLUT3 | |
Polyclonal | |
Unconjugated | |
SLC2A3 | |
AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20527, 25551, 6515 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P11169, P32037, Q07647 | |
SLC2A3 | |
A synthesized peptide derived from human SLC2A3(Accession P11169), corresponding to amino acid residues T462-V496. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction