Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571332 Supplier Invitrogen™ Supplier No. PA571332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLUT3 Polyclonal Antibody for Western Blot

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF1 induced signaling. GLUT3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC2A3
Gene Accession No. P11169
Gene Alias AA408729; AL023014; AL024341; AU040424; C78366; FLJ90380; glucose transporter 3; glucose transporter type 3, brain; Glut3; Glut-3; Slc2a3; solute carrier family 2 (facilitated glucose transporter), member 3; Solute carrier family 2 A3 (neuron glucose transporter); solute carrier family 2 member 3; solute carrier family 2, facilitated glucose transporter member 3; solute carrier family 2, member 2; solute carrier family 2, member 3
Gene Symbols SLC2A3
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human GLUT3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6515
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.