Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut3 Rabbit anti-Human, Mouse, Rat, Clone: 6T3F2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Glut3 |
---|---|
Clone | 6T3F2 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Glut3 Monoclonal antibody specifically detects Glut3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Glut3 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6515 | |
IgG | |
Affinity purified |
6T3F2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Glut3 (P11169). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title