Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT4 Monoclonal Antibody (3G10A3)
GREENER_CHOICE

Catalog No. PIMA517176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517176 Supplier Invitrogen™ Supplier No. MA517176
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17176 targets SLC2A4 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17176 immunogen is purified recombinant fragment of human SLC2A4 (amino acids: 224-353 ) expressed in E. Coli. MA5-17176 detects SLC2A4 which has a predicted molecular weight of approximately 54.8kDa.

This gene is a member of the solute carrier family 2 (facilitated glucose transporter) family and encodes a protein that functions as an insulin-regulated facilitative glucose transporter. In the absence of insulin, this integral membrane protein is sequestered within the cells of muscle and adipose tissue. Within minutes of insulin stimulation, the protein moves to the cell surface and begins to transport glucose across the cell membrane. Mutations in this gene have been associated with noninsulin-dependent diabetes mellitus (NIDDM).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3G10A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Slc2a4
Gene Accession No. P14142, P14672
Gene Alias glucose transporter 4; Glucose transporter 4 insuline-responsive; Glucose transporter 4, insuline-responsive; glucose transporter GLUT4; glucose transporter type 4; glucose transporter type 4, insulin-responsive; GLUT4; GLUT-4; GT2; insulin responsive glucose transporter; insulin-responsive glucose transporter; insulin-responsive glucose transporter type 4; MGC93607; SLC2A4; solute carrier family 2 member 4; solute carrier family 2 (facilitated glucose transporter), member 4; solute carrier family 2 , member 4; solute carrier family 2 member 4; solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols Slc2a4
Host Species Mouse
Immunogen Purified recombinant fragment of human SLC2A4 (amino acids: 224-353 ) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20528, 6517
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.