Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut5 Antibody (195205), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1349PEATT594
Description
Glut5 Monoclonal antibody specifically detects Glut5 in Human samples. It is validated for Flow CytometrySpecifications
Glut5 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6518 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Flow Cytometry | |
195205 | |
Flow Cytometry | |
Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5 | |
NS0 mouse myeloma cell line transfected with human Glut5, Met1-Gln501, Accession # P22732 | |
0.2 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction