Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut5 Antibody (OTI9F3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271285H
Description
Glut5 Monoclonal specifically detects Glut5 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Glut5 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SLC2A5 | |
Full length recombinant protein of human Glut5 (NP_003030) produced in HEK293T cells. | |
0.1 mL | |
6518.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI9F3 | |
Western Blot, Immunohistochemistry | |
Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction