Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLUT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580024
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580024 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580024 Supplier Invitrogen™ Supplier No. PA580024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, rat kidney tissue, mouse testis tissue, mouse kidney tissue. IHC: Rat Kidney Tissue, mouse ggj3249 tissue IHC-F: Rat Intestine Tissue.

Cytochalasin B-sensitive carrier. Seems to function primarily as a fructose transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT5
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene SLC2A5
Gene Accession No. P43427, Q9WV38
Gene Alias AI526984; Fructose transporter; glucose transporter type 5, small intestine; glucose transporter-like protein 5; GLUT5; GLUT-5; RP5-963K15.1; SLC2A5; Slc5a; solute carrier family 2 (facilitated glucose transporter), member 5; solute carrier family 2 (facilitated glucose/fructose transporter), member 5; solute carrier family 2 member 5; solute carrier family 2, facilitated glucose transporter member 5; solute carrier family 2, member 5; testicular tissue protein Li 81
Gene Symbols SLC2A5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of mouse SLC2A5 (232-251aa ALQTLRGWKDVHLEMEEIRK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56485, 65197
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.