Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-1R Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 19 publications

$243.50 - $499.50

Specifications

Antigen GLP-1R
Concentration 1.0 mg/mL
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 - 1:400, Dot Blot reported in scientific literature (PMID 27435156), KnockDown Validated reported in scientific literature (PMID 31900217)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, KnockDown
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP197308SS
SDP
View Documents
Novus Biologicals
NBP197308SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NBP197308
SDP
View Documents
Novus Biologicals
NBP197308
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

GLP-1R Polyclonal specifically detects GLP-1R in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated.
Specifications

Specifications

GLP-1R
Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200 - 1:400, Dot Blot reported in scientific literature (PMID 27435156), KnockDown Validated reported in scientific literature (PMID 31900217)
Polyclonal
Rabbit
Cancer, GPCR, Lipid and Metabolism, Signal Transduction
PBS with 0.02% Sodium Azide
GLP-1 receptor, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331
GLP1R
IgG
Affinity Purified
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, KnockDown
Unconjugated
RUO
Human, Mouse, Rat, Canine
P43220
2740
A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.