Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUT9 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP106565F
Description
GLUT9 Polyclonal specifically detects GLUT9 in Human, Mouse, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GLUT9 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Q9NRM0 | |
SLC2A9 | |
Synthetic peptide made to an internal portion of human GLUT9 (within residues 10-60). [Swiss-Prot# Q9NRM0] | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for the human GLUT9 protein Isoform 1 (SLC2A9-L). | |
Store at 4C in the dark. |
Immunohistochemistry (Paraffin) | |
FITC | |
PBS with 0.05% Sodium Azide | |
facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1 | |
Rabbit | |
58.7 kDa | |
0.1 mL | |
Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
56606 | |
Human, Mouse, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction