Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$192.94 - $517.59

Specifications

Antigen GLUT9
Concentration 1 mg/mL
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry-Paraffin 1:200
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP106271SS
SDP
View Documents
Novus Biologicals
NBP106271SS
0.025 mL
Each for $192.94
Only null left
Add to Cart
 
NBP106271
SDP
View Documents
Novus Biologicals
NBP106271
0.1 mL
Each for $517.59
Only null left
Add to Cart
 
Description

Description

GLUT9 Polyclonal specifically detects GLUT9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

GLUT9
Western Blot 0.5 ug/ml, Immunohistochemistry-Paraffin 1:200
Polyclonal
Rabbit
Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
56606
Synthetic peptide made to an internal portion of mouse GLUT9 (within residues 390-440). [Swiss-Prot# Q7TSK9]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
55 kDa
1 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1
SLC2A9
IgG
Affinity Purified
Mouse. Cross-reactivity with human is very low.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.