Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589925 Supplier Thermo Scientific Supplier No. PA589925
Add to Cart
Add to Cart

GLUT9 Polyclonal Antibody

GLUT9 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC2A9
Gene Accession No. Q3T9X0, Q9NRM0
Gene Alias facilitated glucose transporter, type 9; glucose transporter type 9; GLUT9; GLUT-9; GLUTX; human glucose transporter-like protein-9; Slc2a9; SLC2a9A; SLC2A9B; solute carrier family 2 (facilitated glucose transporter), member 9; solute carrier family 2 member 9; solute carrier family 2, facilitated glucose transporter member 9; UAQTL2; Urate transporter; urate voltage-driven efflux transporter 1; URATv1
Gene Symbols SLC2A9
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 429-529 of human SLC2A9 (NP_0644252).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117591, 56606
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.