Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT9 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502207216
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7216 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7216 Supplier Fabgennix Inc Supplier No. GLUT9901AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLUT9 Polyclonal antibody specifically detects GLUT9 in Human samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen GLUT9
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SLC2A9
Gene Accession No. Q9NRM0
Gene Alias facilitated glucose transporter, type 9; glucose transporter type 9; GLUT9; GLUT-9; GLUTX; human glucose transporter-like protein-9; Slc2a9; SLC2a9A; SLC2A9B; solute carrier family 2 (facilitated glucose transporter), member 9; solute carrier family 2 member 9; solute carrier family 2, facilitated glucose transporter member 9; UAQTL2; Urate transporter; urate voltage-driven efflux transporter 1; URATv1
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 50-570 on Glut 9 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56606
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.