Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Dehydrogenase Antibody (SR1881), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322240100UL
Description
Glutamate Dehydrogenase Monoclonal antibody specifically detects Glutamate Dehydrogenase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
Glutamate Dehydrogenase | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1881 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
EC 1.4.1, EC 1.4.1.3, GDH, GDH 1, GDH1, GLUD, glutamate dehydrogenase (NAD(P)+), glutamate dehydrogenase 1, glutamate dehydrogenase 1, mitochondrial, MGC132003 | |
A synthesized peptide derived from human Glutamate Dehydrogenase (Uniprot #: P00367) | |
100 μg | |
Cellular Markers, Diabetes Research, Mitochondrial Markers, Neuroscience | |
2746 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction