Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glutamate-Glo Assay, 5ml

Catalog No. PRJ7021 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ7021 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ7021 Supplier Promega Supplier No. J7021
Only null left
Add to Cart
Add to Cart

The Glutamate-Glo Assay is a bioluminescent assay for rapid, selective and sensitive detection of glutamate in biological samples. The assay couples glutamate oxidation and NADH production with a bioluminescent NADH detection system.

  • Detect Glutamate from a Variety of Biological Samples
  • Amenable to high−throughput formats
  • Detect even intracellular levels of glutamate
  • Limit of detection in the nM range

Specifications

Quantity 5 mL
Content And Storage <-65°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.