Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Receptor 4 Antibody (SR1025), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.59 - $417.40
Specifications
| Antigen | Glutamate Receptor 4 |
|---|---|
| Clone | SR1025 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
Description
Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
| Glutamate Receptor 4 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4 | |
| A synthesized peptide derived from human Glutamate Receptor 4 (Uniprot #: P48058) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1025 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2893 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title