Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Receptor 4 Rabbit anti-Mouse, Rat, Clone: 4M5D0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Glutamate Receptor 4 |
---|---|
Clone | 4M5D0 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Mouse, Rat samples. It is validated for Western BlotSpecifications
Glutamate Receptor 4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2893 | |
IgG | |
Affinity purified |
4M5D0 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4 | |
A synthetic peptide corresponding to a sequence within amino acids 800-902 of human Glutamate Receptor 4 (P48058). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title