Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Receptor 6 Antibody (8A1F11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Glutamate Receptor 6 |
---|---|
Clone | 8A1F11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61788
![]() |
Novus Biologicals
NBP261788 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Glutamate Receptor 6 Monoclonal specifically detects Glutamate Receptor 6 in Human samples. It is validated for Western Blot, ELISA.Specifications
Glutamate Receptor 6 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
2898 | |
Purified recombinant fragment of human GRIK2 (AA: extra 45-226) expressed in E. Coli. | |
Primary | |
102.5 kDa |
8A1F11 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
GRIK2 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title