Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Receptor 6 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298590APC
Description
Glutamate Receptor 6 Polyclonal antibody specifically detects Glutamate Receptor 6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Glutamate Receptor 6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
Produced in rabbits immunized with E. coli-derived Human Glutamate Receptor 6 fragment. | |
0.1 mL | |
Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction | |
2898 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction