Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamate Receptor 6 Rabbit anti-Human, Mouse, Rat, Clone: 4B4L9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316867100UL
This item is not returnable.
View return policy
Description
Glutamate Receptor 6 Monoclonal antibody specifically detects Glutamate Receptor 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Glutamate Receptor 6 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4B4L9 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427 | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Glutamate Receptor 6 (Q13002). | |
100 μg | |
Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction | |
2898 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction