Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ GlutaMAX™ Supplement
GREENER_CHOICE

Catalog No. a1286001
Encompass_Preferred
Click to view available options
Quantity:
100 mL
1 L
Format:
Bag - BioProcess Container (BPC)
Bottle

CTS™ GlutaMAX™ Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS™ GlutaMAX™ Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

CTS GlutaMAX Supplement is an animal origin free (AOF) dipeptide alternative to L-glutamine, with increased stability that improves cell health. CTS GlutaMAX Supplement can be used as a direct substitute for L-glutamine at equimolar concentrations in mammalian and stem cell culture with minimal or no adaptation.

Gibco CTS products are designed for use in manufacturing of cell or gene products, supporting your transition from research to clinical applications.

Compared to L-glutamine, CTS GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up - Unlike L-glutamine, CTS GlutaMAX Supplement does not spontaneously break down to form ammonia. This prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
  • Improves growth efficiency and performance of mammalian cell cultures - Lower levels of ammonia result in improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of CTS GlutaMAX Supplement that is not possible with L-glutamine.
  • Remains stable across a wide range of temperatures - CTS GlutaMAX Supplement retains full functionality at room temperature or 2 to 8°C, for convenient storage and handling.

Closed systems compatible

This size of CTS GlutaMAX Supplement is provided in bioprocess container format with the chamber constructed from Aegis 5-14 film. Sterile, weldable C-Flex™ tubing with a female luer lock provides versatile connection options for integrating into a closed system manufacturing process.

CTS quality

The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001 to certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage Conditions: Room Temperature
Cell Type Mammalian and stem cell culture
Classification Xeno-free
Concentration 100X
Form Liquid
Product Type Supplement
Sterility Sterile-filtered
Format Bottle
For Use With (Application) Cell and Gene Therapy Research, Development and Manufacturing
Green Features Sustainable packaging
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Room Temperature
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.