Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutaminase Antibody (GT1075), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313512
This item is not returnable.
View return policy
Description
Glutaminase Monoclonal antibody specifically detects Glutaminase in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Glutaminase | |
| Monoclonal | |
| Unconjugated | |
| AAD20, EC 3.5.1.2, GLS1DKFZp686O15119, glutaminase, glutaminase C, glutaminase kidney isoform, mitochondrial, glutaminase, phosphate-activated, K-glutaminase, KIAA0838FLJ10358, L-glutamine amidohydrolase | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutaminase (GAC). The exact sequence is proprietary. | |
| 100 μg | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 2744 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunofluorescence | |
| GT1075 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction