Learn More
Invitrogen™ Glutaminase C (GAC) Polyclonal Antibody

Description
This antibody specifically recognizes GAC but not the KGA isoform. Predicted reactivity: Bovine (100%). Recommended positive controls: MDA-MB-231 cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Glutaminase C (GAC) |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.96 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | GLS |
| Gene Accession No. | O94925 |
| Gene Alias | 6330442B14; AAD20; AI314027; B230365M23Rik; GAC; GAM; GLS; Gls1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; RATGLUT |
| Gene Symbols | GLS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.