Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutaminase Monoclonal Antibody (9G6)
GREENER_CHOICE

Catalog No. PIMA549191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549191 Supplier Invitrogen™ Supplier No. MA549191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human HEL whole cell, rat brain tissue, mouse brain tissue. IHC: human gall bladder adenosquamous carcinoma tissue, human appendiceal adenocarcinoma tissue, human liver cancer tissue, human placenta tissue. ICC/IF: SiHa cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Glutaminase is a phosphate-activated amidohydrolase that catalyzes the hydrolysis of glutamine to glutamate and ammonia. It is primarily expressed in the brain and kidney and plays an essential role in generating energy for metabolism, synthesizing neurotransmitter glutamate, and maintaing acid-based balance in the kidney. There are alternate splicing results in multiple transcript variants in the Glutaminase gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutaminase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 9G6
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GLS
Gene Accession No. D3Z7P3, O94925, P13264
Gene Alias 6330442B14; AAD20; AI314027; B230365M23Rik; DKFZp686O15119; FLJ10358; GA; GAC; GAM; GLS; GLS1; Glut; glutaminase; glutaminase C; glutaminase kidney isoform, mitochondrial; Glutaminase kidney isoform, mitochondrial 65 kDa chain; Glutaminase kidney isoform, mitochondrial 68 kDa chain; glutaminase, phosphate-activated; KGA; K-glutaminase; KIAA0838; L-glutamine amidohydrolase; PAG; phosphate-activated glutaminase; RATGLUT
Gene Symbols GLS
Host Species Mouse
Immunogen E.coli-derived human Glutaminase/GLS recombinant protein (Position: K396-N654).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14660, 24398, 2744
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.