Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Glutamine/Glutamate-Glo Assay, 50ml

Catalog No. PRJ8022 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRJ8022 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRJ8022 Supplier Promega Supplier No. J8022
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Glutamine/Glutamate-Glo Assay is a bioluminescent assay for rapid, selective and sensitive detection of glutamine and glutamate in biological samples. The assay is based on the conversion of glutamine to glutamate by Glutaminase enzyme.

  • Sensitive, Quick Detection of Glutamine and Glutamate in Biological Samples
  • Detect even small changes in metabolites
  • Multiplex with other metabolite or viability assays
  • Perform on a variety of sample types

Specifications

Quantity 50 mL
Content And Storage <-65°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.