Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutamine Synthetase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$192.94 - $519.67
Specifications
| Antigen | Glutamine Synthetase |
|---|---|
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:12000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11041404T
![]() |
Novus Biologicals
NB11041404SS |
0.025 mL |
Each for $192.94
|
|
|||||
NB11041404
![]() |
Novus Biologicals
NB11041404 |
0.1 mL |
Each for $519.67
|
|
|||||
Description
Glutamine Synthetase Polyclonal specifically detects Glutamine Synthetase in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Glutamine Synthetase | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| Whole antisera with 0.05% Sodium Azide | |
| 2752 | |
| Recombinant Glutamine Synthetase. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 1:5000, Immunohistochemistry 1:12000, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Bovine | |
| cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43 | |
| GLUL | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title