Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glutamine Synthetase Monoclonal Antibody (CL2999)
GREENER_CHOICE

Catalog No. PIMA535005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535005 Supplier Invitrogen™ Supplier No. MA535005
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: LQSEGSNSDM YLVPAAMFRD PFRKDPNKLV LCEVFKYNRR PAETNLRHTC KRIMDMVSNQ HPWFGMEQEY TLMGTDGHPF GWPSNGFPGP QGPYYCGVGA DRAYGRDIVE AHYRACLYAG VKIAGTNAEV MPAQWEFQIG P Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 92%.

Glutamine synthase is part of the glutamine synthetase family. Ammonia incorporation in animals occurs through the actions of glutamate dehydrogenase and glutamine synthase. Glutamate plays the central role in mammalian nitrogen flow, serving as both a nitrogen donor and nitrogen acceptor. It also has an important role in controlling metabolic regulations of neurotransmitter glutamate. Because of the multiple functions and importance of GS in cellular metabolism, both catalytic activities and synthesis are highly regulated. The activity of GS is controlled by adenylylation. Its activity is decreased in the cerebral cortex of brains affected by Alzheimer's disease, particularly in the vicinity of senile plaques. It is also decreased under conditions of glucose deprivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2999
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLUL
Gene Accession No. P09606, P15104, P15105
Gene Alias cell proliferation-inducing protein 59; GLNA; GLNS; Glul; GLUL protein; Glutamate ammonia ligase; glutamate decarboxylase; glutamate-ammonia ligase; glutamate--ammonia ligase; glutamate-ammonia ligase (glutamine synthase); glutamate-ammonia ligase (glutamine synthetase); glutamine synthase; glutamine synthetase; Glutamine synthetase (glutamate-ammonia ligase); glutamine synthetase 1; glutamine synthetase I; GS; Palmitoyltransferase GLUL; PIG43; PIG59; Proliferation inducing protein 43; proliferation-inducing protein 43
Gene Symbols GLUL
Host Species Mouse
Immunogen Recombinant protein fragment of Human GLUL (Product #RP-88685).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14645, 24957, 2752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.