Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glutamine Synthetase, Mouse, Unlabeled, Clone: 11A3.1,

Catalog No. MABN1182MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1182MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1182MI Supplier MilliporeSigma™ Supplier No. MABN1182
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamine synthetase (EC 4.1.1.15, EC 6.3.1.2; UniProt P09606; also known as Glutamate-ammonia ligase, Glutamate decarboxylase, GS) is encoded by the Glul (also known as Glns) gene (Gene ID 24957) in rat species. Glutamine synthetase (GS) plays an essential role in nitrogen metabolism by catalyzing ATP-dependent condensation of glutamate and ammonia to form glutamine. Three classes of GS exist, class I enzymes (GSI) are specific to prokaryotes and exist as oligomers of 12 identical subunits, class II enzymes (GSII) exist as decamer of identical subunits in eukaryotes and in some bacteria, while class III enzymes (GSIII) function as a double-ringed dodecamer of identical chains found in bacteroides fragilis and in butyrivibrio fibrisolvens. GS is present predominantly in the brain, kidneys, and liver in mammals. In the brain, GS is found primarily in astrocytes, where it participates in the the detoxification of ammonia via assimilation and termination of neurotransmitter signals via recyclization. c-Myc promotes glutamine usage by upregulating glutaminase (GLS), which converts glutamine to glutamate in the TCA cycle, while in some cancer cells Myc-mediated thymine DNA glycosylase (TDG) transcription leads to GS upregulation due to TDG-dependent demethylation of the GS promoter. GS upregulation promotes cancer cell survival under glutamine limitation, while GS silencing decreases cancer cell proliferation and xenograft tumor growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Western Blot
Classification Monoclonal
Clone 11A3.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4) 150mM NaCl with 0.05% Sodium Azide.
Gene Glul, Glns
Gene Accession No. P09606
Host Species Mouse
Immunogen GST-tagged recombinant rat glutamine synthetase internal fragment.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_058769
Test Specificity Clone 11A3.1 targets an internal epitope within the C-terminal half of human and rat glutamine synthetase.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.