Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamyl Prolyl tRNA synthetase/ProRS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982858
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2858 Supplier Bioss Supplier No. BS13393R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. EPRS is a multifunctional aminoacyl-tRNA synthetase that catalyzes the aminoacylation of glutamic acid and proline tRNA species.

Specifications

Antigen Glutamyl Prolyl tRNA synthetase/ProRS
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPRS1
Gene Accession No. P07814
Gene Alias 2410081F06Rik; 3010002K18Rik; Bifunctional aminoacyl-tRNA synthetase; Bifunctional glutamate/proline--tRNA ligase; C79379; Cell proliferation-inducing gene 32 protein; EARS; EPRS; GLNS; GLUPRORS; GluRS; glutamate tRNA ligase; Glutamate--tRNA ligase; Glutamatyl-prolyl-tRNA synthetase; glutamine-proline-tRNA synthetase; glutaminyl-tRNA synthetase; glutamyl-prolyl tRNA synthetase; glutamyl-prolyl-tRNA synthetase; Glutamyl-tRNA synthetase; PARS; PIG32; proliferation-inducing gene 32 protein; proliferation-inducing protein 32; proline tRNA ligase; proline-tRNA ligase; Proline--tRNA ligase; Prolyl-tRNA synthetase; ProRS; QARS; QPRS
Gene Symbols EPRS1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Glutamyl Prolyl tRNA syntase/ProRS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2058
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.