Learn More
Pointe Scientific γ-Glutamyl Transferase Reagents for Hitachi™ Analyzers
Shop All Pointe Scientific ProductsDescription
Includes
Reagent 1 and Reagent 2 when combined: Tris buffer (pH 8.1±0.1) 89mM; Glycylglycine 126mM; GLUPA-C 3.2mM; Sodium azide 0.095%; stabilizer
- Method: Modified Szasz, Kinetic
- Format: Liquid (ready to use)
- Linearity: 800 U/L
- Expected Values: 6 to 54 U/L
Applications:
For the quantitative kinetic determination of γ-glutamyl transferase (γ-GT) activity in serum on Hitachi analyzers
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | gamma-Glutamyl Transferase R1 Kit |
| For Use With (Application) | Quantitative kinetic determination of gamma Glutamyl Transferase activity in Serum |
| Quantity | 3 x 410 R1mL |
| Form | Liquid |
For use with in vitro diagnostics only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.