Glutaraldehyde, 25% aq. soln.

Manufacturer: Alfa Aesar A17876AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA17876AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 111-30-8
    Molecular Formula OHC(CH2)3CHO
    Linear Formula OHC(CH2)3CHO
    Formula Weight 100.12
    MDL Number MFCD00007025
    UN Number UN2922
    Beilstein 605390
    Merck Index 14,4472
    Chemical Name or Material Glutaraldehyde

    Specifications

    Formula Weight 100.12
    Quantity 500mL
    TSCA TSCA
    Linear Formula OHC(CH2)3CHO
    MDL Number MFCD00007025
    UN Number UN2922
    Beilstein 605390
    Merck Index 14,4472
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H334-H314-H302-H317
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause an allergic skin
    Health Hazard 3 GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYE
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    RTECS Number MA2450000
    Density 1.062
    Assay Percent Range 0.25
    Molecular Formula OHC(CH2)3CHO
    CAS 111-30-8
    Chemical Name or Material Glutaraldehyde

    Description

    Applications
    Glutaraldehyde is used for cross-linking proteins and polyhydroxy materials. Also used as a fixative for tissues and as an amine-reactive homobifunctional crosslinker and as a disinfectant for medical equipment. It is also used in coupling reaction of biotin hydrazides to nucleic acids which is useful for conjugating fluorescent hydrazides and hydroxylamines to DNA and for stabilization ofproteins on agarose beads,activation of polystyrene and glass, for immobilization of antibodies and antigens. It is a disinfectant, which is rapidly effective against vegetative forms of Gram-positiveand Gram-negative bacteria.

    Solubility
    Miscible with water, ethanol, benzene, ether, acetone, dichloromethane, ethylacetate, isopropanol, n-hexane and toluene.

    Notes
    Heat and air sensitive. Incompatible with strong acids, strong bases and strong oxidizing agents.

    GHS H Statement
    H331-H334-H314-H302-H317
    Toxic if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    May cause an allergic skin

    2° to 8°C

    GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYE

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit