Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutaraldehyde, 25% Aqueous Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Specifications
Specifications
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 4 L |
Specific Gravity | 1L = 1.09kg |
Formula Weight | 100.12 |
Percent Purity | 25% aq. soln. |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Glutaraldehyde |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction