Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaraldehyde, 25% in H2O, Practical, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

111-30-8
C5H8O2
100.117
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
CHEBI:64276
Quantity 4L
CAS 111-30-8
Molecular Formula C5H8O2
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
SMILES C(CC=O)CC=O
IUPAC Name pentanedial
Molecular Weight (g/mol) 100.117
PubChem CID 3485
ChEBI CHEBI:64276
Formula Weight 100.12
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.09kg
Chemical Name or Material Glutaraldehyde
Assay 25% in H2O
Grade Practical Grade
Show More Show Less