Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Glutaraldehyde Solution, 25%
SCI_ED

Catalog No. s25341 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
Packaging:
25%
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 111-30-8, 7732-18-5
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00007025
InChI Key SXRSQZLOMIGNAQ-UHFFFAOYSA-N
Synonym glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
PubChem CID 3485
ChEBI CHEBI:64276
IUPAC Name pentanedial
SMILES O=CCCCC=O

Specifications

Density 1.06g/mL
Assay Percent Range 25% w/v
Packaging 25%
Quantity 1 L
Formula Weight 100.2
Chemical Name or Material Glutaraldehyde Solution, 25%
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.