Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutaredoxin 3/GLRX3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220766
Description
Glutaredoxin 3/GLRX3 Polyclonal specifically detects Glutaredoxin 3/GLRX3 in Human samples. It is validated for Western Blot.Specifications
TXNL2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
FLJ11864, GLRX4, glutaredoxin 3, glutaredoxin 4, glutaredoxin-3, GRX3, GRX4, PICOTbA500G10.4, PKC-interacting cousin of thioredoxin, PKCq-interacting protein, PKC-theta-interacting protein, thioredoxin-like 2, Thioredoxin-like protein 2, TXNL2, TXNL3 | |
Rabbit | |
37 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (92%), Rat (92%), Xenopus laevis (80%), Bovine (93%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.38mg/mL | |
Western Blot 1:500-1:3000 | |
O76003 | |
GLRX3 | |
Recombinant protein encompassing a sequence within the center region of human TXNL2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
10539 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction