Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaredoxin 5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982860
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2860 Supplier Bioss Supplier No. BS13395R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLRX5 is monothiol glutaredoxin involved in the biogenesis of iron-sulfur clusters. GLRX5 required for normal iron homeostasis. GLRX5 is required for normal regulation of hemoglobin synthesis by the iron-sulfur protein ACO1.

Specifications

Antigen Glutaredoxin 5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GLRX5
Gene Accession No. Q86SX6
Gene Alias C14orf87; FLB4739; GLRX5; glutaredoxin 5; glutaredoxin 5 homolog; Glutaredoxin-related protein 5, mitochondrial; GRX5; Monothiol glutaredoxin-5; PR01238; PRO1238; PRSA; SIDBA3; SPAHGC
Gene Symbols GLRX5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Glutaredoxin 5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51218
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.