Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaric anhydride, 98%

Catalog No. AAA111520B
Click to view available options
Quantity:
50 g
100 g
250 g
1000 g

CAS: 108-55-4 | C5H6O3 | 114.1 g/mol

Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaric anhydride is used in the production of glutaric acid family pharmaceuticals. Its derivatives are also used in certain corrosion inhibitors, some surfactants and metal finishing compounds. It is also used in the production of polymers, particularly modified unsaturated linear (epoxy) polyesters and as a raw material in the manufacture of flavors. It is also used in improvement of electrode/electrolyte interfaces in High-Voltage Spinel Lithium-Ion batteries as an electrolyte additive.

Solubility
Soluble in Ether, dimethyl sulfoxide and methanol and water.

Notes
Moisture sensitive. Incompatible withstrong oxidizing agents, acids, bases, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-55-4
Molecular Formula C5H6O3
Molecular Weight (g/mol) 114.1
MDL Number MFCD00006679
InChI Key VANNPISTIUFMLH-UHFFFAOYSA-N
Synonym glutaric anhydride, dihydro-2h-pyran-2,6 3h-dione, glutaric acid anhydride, pentanedioic anhydride, pyroglutaric acid, 2h-pyran-2,6 3h-dione, dihydro, pentanedioic acid anhydride, anhydrid kyseliny glutarove, unii-63ofi15s80, anhydrid kyseliny glutarove czech
PubChem CID 7940
IUPAC Name oxane-2,6-dione
SMILES C1CC(=O)OC(=O)C1

Specifications

Melting Point 52°C to 57°C
Density 1.411
Boiling Point 291°C
Flash Point 178°C (352°F)
Assay Percent Range 98%
Quantity 1000 g
Beilstein 110051
Sensitivity Moisture sensitive
Solubility Information Soluble in Ether,dimethyl sulfoxide and methanol and water.
Formula Weight 114.1
Percent Purity 98%
Chemical Name or Material Glutaric anhydride
Show More Show Less
Hazard Category H302-H312-H315-H318
Hazard Statement GHS H Statement
H318-H312-H335-H315
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 203-593-6
RTECSNumber MA3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.