Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutarimide, 98%

Catalog No. aal0096806
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1121-89-7 | C5H7NO2 | 113.12 g/mol

Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutarimide acts as an inhibitor of protein synthesis. Further, it is used as a reactant for thionations and biocatalytic asymmetric synthesis of sitagliptin production. It is also employed in the generation of beta-adrenoceptor ligands, enantioselective synthesis of securinega alkaloids and alfa-fluoro-alfa amino amides. In addition to this, it is used in intramolecular amidocyclopropanation reactions.

Solubility
Soluble in water, hot ethanol and boiling benzene. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-89-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00006670
InChI Key KNCYXPMJDCCGSJ-UHFFFAOYSA-N
Synonym glutarimide, 2,6-piperidinedione, 2,6-diketopiperidine, 2,6-dioxopiperidine, acetylglycinamide impurity b, piperidin-2,6-dione, acmc-2099da, 5-21-09-00557 beilstein handbook reference, 2,6-piperidinedione 9ci
PubChem CID 70726
ChEBI CHEBI:5435
IUPAC Name piperidine-2,6-dione
SMILES O=C1CCCC(=O)N1

Specifications

Melting Point 153°C to 157°C
Odor Nearly Odorless
Quantity 5 g
Beilstein 110052
Solubility Information Soluble in water,hot ethanol and boiling benzene. Insoluble in ether.
Formula Weight 113.12
Percent Purity 98%
Chemical Name or Material Glutarimide
EINECSNumber 214-340-4
RTECSNumber MA4000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.