Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 1/GPX1 Antibody (3G1), Novus Biologicals™
SDP

Catalog No. p-200022390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297884 50 μL
NB297885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297884 Supplier Novus Biologicals Supplier No. NBP32654650UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glutathione Peroxidase 1/GPX1 Monoclonal antibody specifically detects Glutathione Peroxidase 1/GPX1 in Human samples. It is validated for ELISA

Specifications

Antigen Glutathione Peroxidase 1/GPX1
Applications ELISA
Classification Monoclonal
Clone 3G1
Conjugate Unconjugated
Dilution ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Glutathione Peroxidase 1/GPX1 [UniProt P07203]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2876
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.