Learn More
Glutathione Peroxidase 2/GPX2 Antibody (HL125), Novus Biologicals™
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP313662
Catalog No. NB177349
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Glutathione Peroxidase 2/GPX2 Monoclonal antibody specifically detects Glutathione Peroxidase 2/GPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
Glutathione Peroxidase 2/GPX2
Monoclonal
Unconjugated
EC 1.11.1, EC 1.11.1.9, Gastrointestinal glutathione peroxidase, gastrointestinal glutathione peroxidase 2, GI-GPx, glutathione peroxidase 2, glutathione peroxidase 2 (gastrointestinal), Glutathione peroxidase-gastrointestinal, Glutathione peroxidase-related protein 2, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPx-2, GSHPx-GI
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Glutathione Peroxidase 2/GPX2. The exact sequence is proprietary.
100 μg
Cell Biology, Endocrinology
2877
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
HL125
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.