Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 2/GPX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB092928 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB092928 20 μL
NB092927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB092928 Supplier Novus Biologicals Supplier No. NBP30320920UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glutathione Peroxidase 2/GPX2 Polyclonal specifically detects Glutathione Peroxidase 2/GPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glutathione Peroxidase 2/GPX2
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 1.11.1, EC 1.11.1.9, Gastrointestinal glutathione peroxidase, gastrointestinal glutathione peroxidase 2, GI-GPx, glutathione peroxidase 2, glutathione peroxidase 2 (gastrointestinal), Glutathione peroxidase-gastrointestinal, Glutathione peroxidase-related protein 2, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPx-2, GSHPx-GI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-180 of human Glutathione Peroxidase 2/GPX2 (NP_002074.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2877
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.