Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione Peroxidase 4/GPX4 Antibody (9D4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Glutathione Peroxidase 4/GPX4 |
---|---|
Clone | 9D4 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Glutathione Peroxidase 4/GPX4 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase | |
A synthesized peptide derived from Human Glutathione Peroxidase 4/GPX4 [UniProt P36969] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9D4 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2879 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title