Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione Peroxidase 4/GPX4 Antibody (LHM 2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308253MFV610
Description
Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Glutathione Peroxidase 4/GPX4 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
LHM 2 | |
Flow Cytometry | |
EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase | |
A 375P cells crude extract | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
2879 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction