Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 4 Mouse anti-Mouse, Human, Unconjugated, Clone: Mab63-1, MilliporeSigma™

Catalog No. MABF196925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1969-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1969-25 Supplier MilliporeSigma Supplier No. MABF196925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Glutathione Peroxidase 4, clone Mab63-1, Cat. No. MABF1969, is a mouse monoclonal antibody that detects cytosolic glutathione peroxidase 4 and has been tested for use in Dot Blot, Electron Microscopy, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione Peroxidase 4
Applications Dot Blot, Immunomicroscopy, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Mab63-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Phospholipid hydroperoxide glutathione peroxidase cytosolic;cGPx4;PHGPx;Glutathione peroxidase 4;GPx-4;GSHPx-4
Gene Symbols GPX4
Host Species Mouse
Immunogen His-tagged purified recombinant human cytosolic glutathione peroxidase 4.
Molecular Weight of Antigen ∽19 kDa observed; 19.53 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone Mab63-1 specifically detects cytosolic Glutathione Peroxidase 4 in human and murine cells. it targets a minimum epitope of 6 amino acids from the N-terminal region.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.