Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Reductase Monoclonal Antibody (2A4-1G9), QED Bioscience™
SDP

Catalog No. 502277549
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7549 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7549 Supplier Leinco Technologies Inc Supplier No. 13902200UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutathione Reductase Monoclonal antibody specifically detects Glutathione Reductase in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen Glutathione Reductase
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A4-1G9
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Gsr
Gene Accession No. P00390
Gene Alias AI325518; D8Ertd238e; epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; GLUR; glutathione reductase; glutathione reductase 1; glutathione reductase, mitochondrial; glutathione S-reductase; glutathione-disulfide reductase; GR; Gr1; Gr-1; GRase; GRD1; GSR; HEL-75; HEL-S-122m; hypothetical protein LOC553575; zgc:110010
Host Species Mouse
Immunogen Recombinant human glutathione reductase purified from E. coli expressing the human glutathione reductase gene (aa 44-522).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2936
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.