Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S-Transferase/GST Antibody (4D12.H7.D10.E3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2441820.05MG
Description
Glutathione S-Transferase/GST Monoclonal specifically detects Glutathione S-Transferase/GST in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblotting.Specifications
Glutathione S-Transferase/GST | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:1000, ELISA 1:50000 - 1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunoblotting | |
Mouse | |
0.05 mg | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunoblot | |
4D12.H7.D10.E3 | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
This Protein A purified antibody was prepared by repeated immunizations in mice with Glutathione S-Transferase/GSTProtein | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction