Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione-S-transferase (GST) Polyclonal Antibody, QED Bioscience™

Rabbit Polyclonal Antibody
Supplier: Leinco Technologies Inc 188441MG

This item is not returnable.
View return policy
Description
Glutathione-S-transferase (GST) Polyclonal antibody specifically detects Glutathione-S-transferase (GST) in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
Glutathione-S-transferase (GST) | |
Polyclonal | |
Unconjugated | |
activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
Glutathione-S-Transferase (GST) from Schistosoma japonicum. | |
1 mg | |
Primary | |
Antibody | |
Tag |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Rabbit | |
Protein A | |
RUO | |
2-8°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction