Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S Transferase kappa 1 Antibody (16A11.E4.E3.E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2441720.05MG
Description
Glutathione S Transferase kappa 1 Monoclonal specifically detects Glutathione S Transferase kappa 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page.Specifications
Glutathione S Transferase kappa 1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:1000, ELISA 1:50000 - 1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, SDS-Page 1:100-1:2000 | |
EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1 | |
Mouse | |
0.05 mg | |
373156 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
16A11.E4.E3.E7 | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
GSTK1 | |
This Protein A purified antibody was prepared by repeated immunizations in mice with Glutathione S Transferase kappa 1 Protein. | |
Primary | |
Human | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction